missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Ethyl-3-indoleethanol 98.0+%, TCI America™
Supplier: TCI America E082425G
Specifications
7-Ethyl-3-indoleethanol | |
50°C | |
C12H15NO | |
25 g | |
7-Ethyl-3-(2-hydroxyethyl)indole, 7-Ethyltryptophol | |
CCC1=C2NC=C(CCO)C2=CC=C1 | |
189.26 | |
189.26 | |
Crystalline Powder |
41340-36-7 | |
White-Yellow | |
MFCD01718805 | |
3077 | |
UVSDNCAZVSQJQA-UHFFFAOYSA-N | |
2-(7-ethyl-1H-indol-3-yl)ethan-1-ol | |
198231 | |
≥98.0% (GC) |
Chemical Identifiers
41340-36-7 | |
189.26 | |
UVSDNCAZVSQJQA-UHFFFAOYSA-N | |
198231 | |
CCC1=C2NC=C(CCO)C2=CC=C1 |
C12H15NO | |
MFCD01718805 | |
7-Ethyl-3-(2-hydroxyethyl)indole, 7-Ethyltryptophol | |
2-(7-ethyl-1H-indol-3-yl)ethan-1-ol |
Safety and Handling
RTECSNumber : NL8512200
TSCA : No