missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Ethyl-10-hydroxycamptothecin, TCI America™

Manufacturer:  TCI America E07481G

 View more versions of this product

Catalog No. E07481G



Specifications

Specifications

7-Ethyl-10-hydroxycamptothecin
217°C
86639-52-3
MFCD06762720
7-ethyl-10-hydroxycamptothecin, sn 38 lactone, 7-ethyl-10-hydroxy-camptothecin, 10-hydroxy-7-ethylcamptothecin, 7-ethyl-10-hydroxycamptothecine, le-sn38, 7-ethyl-10-hydroxy-20 s-camptothecin, captothecin, 7-ethyl-10-hydroxy, s-4,11-diethyl-4,9-dihydroxy-1h-pyrano 3',4':6,7 indolizino 1,2-b quinoline-3,14 4h,12h-dione, 7-ethyl-10-hydroxy camptothecin
CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)O
104842
392.41
≥98.0% (HPLC)
Yellow
1g
C22H20N2O5
1544
FJHBVJOVLFPMQE-QFIPXVFZSA-N
392.411
CHEBI:8988
Crystal-Powder at 20°C

Chemical Identifiers

86639-52-3
392.411
FJHBVJOVLFPMQE-QFIPXVFZSA-N
104842
CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)O
C22H20N2O5
MFCD06762720
7-ethyl-10-hydroxycamptothecin, sn 38 lactone, 7-ethyl-10-hydroxy-camptothecin, 10-hydroxy-7-ethylcamptothecin, 7-ethyl-10-hydroxycamptothecine, le-sn38, 7-ethyl-10-hydroxy-20 s-camptothecin, captothecin, 7-ethyl-10-hydroxy, s-4,11-diethyl-4,9-dihydroxy-1h-pyrano 3',4':6,7 indolizino 1,2-b quinoline-3,14 4h,12h-dione, 7-ethyl-10-hydroxy camptothecin
CHEBI:8988
Safety and Handling

Safety and Handling

RTECSNumber : UQ0491000

TSCA : No

Recommended Storage : Freezer

Documents