Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Ethoxyresorufin

Fluorimetric substrate for detection of cytochrome P450 linked enzymes

Chemical Identifiers

Chemical Name or Material 7-Ethoxyresorufin
CAS 5725-91-7
Formula Weight 241.24
MDL Number MFCD00037661
Beilstein 225973
Synonym Resorufin ethyl ether; 7-Ethoxy-3H-phenoxazin-3-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ62987LB0

  SDS
alfa aesar™
J62987LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 241.24
Melting Point 237° to 240°C
Chemical Name or Material 7-Ethoxyresorufin
Physical Form Powder
MDL Number MFCD00037661
CAS 5725-91-7
Beilstein 225973
Synonym Resorufin ethyl ether; 7-Ethoxy-3H-phenoxazin-3-one
Storage Note 1 2° to 8°C
Color Brown
Quantity 5mg
Safety and Handling

2° to 8°C