missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Ethoxycoumarin, TCI America™

Manufacturer:  TCI America E053825G

 View more versions of this product

Catalog No. E053825G



Specifications

Specifications

7-Ethoxycoumarin
90°C
31005-02-4
MFCD00006877
LIFAQMGORKPVDH-UHFFFAOYSA-N
7-ethoxychromen-2-one
35703
190.20
≥97.0% (GC)
Undesignated
25g
C11H10O3
7-ethoxycoumarin, 7-ethoxy-2h-chromen-2-one, ethylumbelliferone, 2h-1-benzopyran-2-one, 7-ethoxy, 7-ethoxy-2h-1-benzopyran-2-one, 7-ethoxycoumarine, 7-ethoxycoumarin, pubchem8680, coumarin, 7-ethoxy, acmc-209hjx
CCOC1=CC2=C(C=C1)C=CC(=O)O2
190.198
CHEBI:28184
Crystal-Powder at 20°C

Chemical Identifiers

31005-02-4
190.198
LIFAQMGORKPVDH-UHFFFAOYSA-N
35703
7-ethoxychromen-2-one
C11H10O3
MFCD00006877
7-ethoxycoumarin, 7-ethoxy-2h-chromen-2-one, ethylumbelliferone, 2h-1-benzopyran-2-one, 7-ethoxy, 7-ethoxy-2h-1-benzopyran-2-one, 7-ethoxycoumarine, 7-ethoxycoumarin, pubchem8680, coumarin, 7-ethoxy, acmc-209hjx
CHEBI:28184
CCOC1=CC2=C(C=C1)C=CC(=O)O2
Safety and Handling

Safety and Handling

TSCA : No

Documents