Please login to your online account to display your discounted pricing

7-Ethoxycoumarin, 99.5%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  296310010

 View more versions of this product

Catalog No. AC296310010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Color Beige
Melting Point 87° to 90°C
Quantity 1g
Chemical Name or Material 7-Ethoxycoumarin
Name Note 99.5%
CAS 31005-02-4
CAS Min % 99.4
CAS Max % 100.0
Physical Form Beige crystalline powder
Assay 99.4% min.
Assay Percent Range 99.50%
Molecular Formula C11H10O3
Formula Weight 190.2g/mol
MDL Number MFCD00006877
Beilstein 18, 28
EINECS Number 250-429-4

Specifications

Chemical Name or Material 7-Ethoxycoumarin
Name Note 99.5%
CAS 31005-02-4
CAS Min % 99.4
CAS Max % 100.0
Assay 99.4% min.
Assay Percent Range 99.50%
Molecular Formula C11H10O3
Formula Weight 190.2g/mol
MDL Number MFCD00006877