Please login to your online account to display your discounted pricing

7-Ethoxycoumarin, 99.5%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 7-Ethoxycoumarin
Name Note 99.5%
CAS 31005-02-4
CAS Min % 99.4
CAS Max % 100.0
Assay 99.4% min.
Assay Percent Range 99.50%
Molecular Formula C11H10O3
Formula Weight 190.2g/mol
MDL Number MFCD00006877
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC296310010

  SDS
thermo fisher scientific
296310010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Color Beige
Melting Point 87° to 90°C
Quantity 1g
Chemical Name or Material 7-Ethoxycoumarin
Name Note 99.5%
CAS 31005-02-4
CAS Min % 99.4
CAS Max % 100.0
Physical Form Beige crystalline powder
Assay 99.4% min.
Assay Percent Range 99.50%
Molecular Formula C11H10O3
Formula Weight 190.2g/mol
MDL Number MFCD00006877
Beilstein 18, 28
EINECS Number 250-429-4