missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Ethoxycoumarin, 99%, Thermo Scientific Chemicals
$85.22 - $262.58
Chemical Identifiers
CAS | 31005-02-4 |
---|---|
Molecular Formula | C11H10O3 |
Molecular Weight (g/mol) | 190.198 |
MDL Number | MFCD00006877 |
InChI Key | LIFAQMGORKPVDH-UHFFFAOYSA-N |
Synonym | 7-ethoxycoumarin, 7-ethoxy-2h-chromen-2-one, ethylumbelliferone, 2h-1-benzopyran-2-one, 7-ethoxy, 7-ethoxy-2h-1-benzopyran-2-one, 7-ethoxycoumarine, pubchem8680, coumarin, 7-ethoxy, acmc-209hjx |
PubChem CID | 35703 |
ChEBI | CHEBI:28184 |
IUPAC Name | 7-ethoxychromen-2-one |
SMILES | CCOC1=CC2=C(C=C1)C=CC(=O)O2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL14116MC
|
Thermo Scientific Chemicals
L14116MC |
100 mg |
Each for $85.22
|
|
|||||
AAL14116ME
|
Thermo Scientific Chemicals
L14116ME |
500 mg |
Each for $262.58
|
|
|||||
Description
7-Ethoxycoumarin is a substrate for CYP2B6 used in fluorometric assays of microsomal monooxygenase activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications7-Ethoxycoumarin is a substrate for CYP2B6 used in fluorometric assays of microsomal monooxygenase activity.
Solubility
Soluble in hot methanol (very faint turbidity), 95% ethanol (50 mg/ml), DMF, and DMSO. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Chemical Identifiers
31005-02-4 | |
190.198 | |
LIFAQMGORKPVDH-UHFFFAOYSA-N | |
35703 | |
7-ethoxychromen-2-one |
C11H10O3 | |
MFCD00006877 | |
7-ethoxycoumarin, 7-ethoxy-2h-chromen-2-one, ethylumbelliferone, 2h-1-benzopyran-2-one, 7-ethoxy, 7-ethoxy-2h-1-benzopyran-2-one, 7-ethoxycoumarine, pubchem8680, coumarin, 7-ethoxy, acmc-209hjx | |
CHEBI:28184 | |
CCOC1=CC2=C(C=C1)C=CC(=O)O2 |
Specifications
31005-02-4 | |
C11H10O3 | |
100 mg | |
7-ethoxycoumarin, 7-ethoxy-2h-chromen-2-one, ethylumbelliferone, 2h-1-benzopyran-2-one, 7-ethoxy, 7-ethoxy-2h-1-benzopyran-2-one, 7-ethoxycoumarine, pubchem8680, coumarin, 7-ethoxy, acmc-209hjx | |
LIFAQMGORKPVDH-UHFFFAOYSA-N | |
7-ethoxychromen-2-one | |
35703 | |
190.2 | |
7-Ethoxycoumarin |
88°C to 91°C | |
MFCD00006877 | |
146200 | |
Soluble in hot methanol (very faint turbidity),95% ethanol (50mg/ml),DMF,and DMSO. Insoluble in water. | |
CCOC1=CC2=C(C=C1)C=CC(=O)O2 | |
190.198 | |
CHEBI:28184 | |
99% |
Safety and Handling
EINECSNumber : 250-429-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only