Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Ethoxy-4-methylcoumarin, 98%

$47.26 - $175.47

Chemical Identifiers

Chemical Name or Material 7-Ethoxy-4-methylcoumarin
CAS 87-05-8
Molecular Formula C12H12O3
Formula Weight 204.23
MDL Number MFCD00016971
Beilstein 169998
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2006506

 
alfa aesar™
B2006506
5g Each for $47.26

AAB2006514

 
alfa aesar™
B2006514
25g Each for $175.47
Description & Specifications

Specifications

Formula Weight 204.23
Chemical Name or Material 7-Ethoxy-4-methylcoumarin
TSCA TSCA
Assay 98%
Molecular Formula C12H12O3
MDL Number MFCD00016971
CAS 87-05-8
Beilstein 169998
Melting Point 113° to 117°C
Quantity 5g