missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Diethylamino-4-methylcoumarin 98.0+%, TCI America™

Manufacturer:  Tci America M0631500G

 View more versions of this product

Catalog No. M0631500G



Specifications

Specifications

7-Diethylamino-4-methylcoumarin
Undesignated
500g
C14H17NO2
4-methyl-7-diethylamino coumarin, 4-methyl-7-diethylaminocoumarin, 7-diethylamino-4-methyl-2h-chromen-2-one, 7-diethylamino-4-methylcoumarin, blancophor aw, blancophor ffg, coumarin 1, coumarin 460, coumarin 47, hakkol p
CCN(CC)C1=CC2=C(C=C1)C(=CC(=O)O2)C
231.295
CHEBI:51938
Crystal-Powder at 20°C
240°C
72°C
91-44-1
MFCD00006864
AFYCEAFSNDLKSX-UHFFFAOYSA-N
7-(diethylamino)-4-methylchromen-2-one
7050
231.30
≥98.0% (GC,T)

Chemical Identifiers

91-44-1
231.295
AFYCEAFSNDLKSX-UHFFFAOYSA-N
7050
7-(diethylamino)-4-methylchromen-2-one
C14H17NO2
MFCD00006864
4-methyl-7-diethylamino coumarin, 4-methyl-7-diethylaminocoumarin, 7-diethylamino-4-methyl-2h-chromen-2-one, 7-diethylamino-4-methylcoumarin, blancophor aw, blancophor ffg, coumarin 1, coumarin 460, coumarin 47, hakkol p
CHEBI:51938
CCN(CC)C1=CC2=C(C=C1)C(=CC(=O)O2)C
Safety and Handling

Safety and Handling

EINECSNumber : (5)-2696

RTECSNumber : GN6370000

TSCA : Yes

Documents