Learn More
7-Deazahypoxanthine, 97%, Thermo Scientific Chemicals
$123.68 - $523.30
Chemical Identifiers
CAS | 3680-71-5 |
---|---|
Molecular Formula | C6H5N3O |
Molecular Weight (g/mol) | 135.13 |
MDL Number | MFCD08445809 |
InChI Key | FBMZEITWVNHWJW-UHFFFAOYSA-N |
Synonym | 7h-pyrrolo 2,3-d pyrimidin-4-ol, pyrrolo 2,3-d pyrimidin-4-ol, 7-deazahypoxanthine, 3h-pyrrolo 2,3-d pyrimidin-4 7h-one, 4-hydroxypyrrolo 2,3-d pyrimidine, 4-hydroxypyrrolopyrimidine, 1h-pyrrolo 2,3-d pyrimidin-4 7h-one, 3,7-dihydro-4h-pyrrolo 2,3-d pyrimidin-4-one, pyrrolo 2,3-d pyrimidin-4-one |
PubChem CID | 96194 |
IUPAC Name | 1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one |
SMILES | O=C1N=CNC2=C1C=CN2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6453706
|
Thermo Scientific Chemicals
H6453706 |
5 g |
Each for $123.68
|
|
|||||
AAH6453714
|
Thermo Scientific Chemicals
H6453714 |
25 g |
Each for $523.30
|
|
|||||
Description
It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Moisture and light sensitive. Store at -20°C
Chemical Identifiers
3680-71-5 | |
135.13 | |
FBMZEITWVNHWJW-UHFFFAOYSA-N | |
96194 | |
O=C1N=CNC2=C1C=CN2 |
C6H5N3O | |
MFCD08445809 | |
7h-pyrrolo 2,3-d pyrimidin-4-ol, pyrrolo 2,3-d pyrimidin-4-ol, 7-deazahypoxanthine, 3h-pyrrolo 2,3-d pyrimidin-4 7h-one, 4-hydroxypyrrolo 2,3-d pyrimidine, 4-hydroxypyrrolopyrimidine, 1h-pyrrolo 2,3-d pyrimidin-4 7h-one, 3,7-dihydro-4h-pyrrolo 2,3-d pyrimidin-4-one, pyrrolo 2,3-d pyrimidin-4-one | |
1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one |
Specifications
3680-71-5 | |
Brown | |
MFCD08445809 | |
607869 | |
7h-pyrrolo 2,3-d pyrimidin-4-ol, pyrrolo 2,3-d pyrimidin-4-ol, 7-deazahypoxanthine, 3h-pyrrolo 2,3-d pyrimidin-4 7h-one, 4-hydroxypyrrolo 2,3-d pyrimidine, 4-hydroxypyrrolopyrimidine, 1h-pyrrolo 2,3-d pyrimidin-4 7h-one, 3,7-dihydro-4h-pyrrolo 2,3-d pyrimidin-4-one, pyrrolo 2,3-d pyrimidin-4-one | |
FBMZEITWVNHWJW-UHFFFAOYSA-N | |
1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one | |
96194 | |
97% | |
7-Deazahypoxanthine |
345°C to 350°C | |
C6H5N3O | |
5 g | |
Light and moisture sensitive | |
Soluble in dimethyl sulfoxide. | |
O=C1N=CNC2=C1C=CN2 | |
135.13 | |
135.13 | |
Solid |
Safety and Handling
GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P280i-P285-P304+P341-P305+P351+P338-P342+P311-P501c
H319-H334
RTECSNumber : UY9440000
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only