missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Deazaguanine, 97%

Manufacturer:  Alfa Aesar™ H54595MD

 View more versions of this product

Catalog No. AAH54595MD



Description

Description

Applications
Ethidium bromide efficiently intercalates into synthetic duplex DNAs with 7-deazaguanine in place of guanine, but shows none of the fluorescence enhancement usually observed upon intercalation. 7-Deazaadenine-duplexes show about 70% of the expected fluorescence enhancement.

Solubility
Soluble in N,N-Dimethylformamide, Dimethyl sulfoxide.

Notes
Store at room temperature. Keep away from oxidizing agents and acids.
Specifications

Specifications

7-Deazaguanine
7355-55-7
MFCD00079103
2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one
OLAFFPNXVJANFR-UHFFFAOYSA-N
2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
96253
97%
250mg
C6H6N4O
512680
Soluble in N,N-Dimethylformamide,Dimethyl sulfoxide.
C1=CNC2=C1C(=O)N=C(N2)N
150.141
150.14

Chemical Identifiers

7355-55-7
150.141
OLAFFPNXVJANFR-UHFFFAOYSA-N
96253
C1=CNC2=C1C(=O)N=C(N2)N
C6H6N4O
MFCD00079103
2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one
2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents