missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Deazaguanine, 90+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450260250

 View more versions of this product

Catalog No. AC450260250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

7-Deazaguanine
>230.0°C
7355-55-7
C6H6N4O
OLAFFPNXVJANFR-UHFFFAOYSA-N
2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
96253
90+%
Glass bottle
25g
90% min. (HPLC)
2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 7-deazaguanine, gpl8t5zo3m, unii-gpl8t5zo3m
C1=CNC2=C1C(=O)N=C(N2)N
150.14
150.14

Chemical Identifiers

7355-55-7
150.14
2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 7-deazaguanine, gpl8t5zo3m, unii-gpl8t5zo3m
2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
C6H6N4O
OLAFFPNXVJANFR-UHFFFAOYSA-N
96253
C1=CNC2=C1C(=O)N=C(N2)N
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

Documents