missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Deazaguanine, 90+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC450260050
Specifications
7-Deazaguanine | |
>230.0°C | |
7355-55-7 | |
C6H6N4O | |
OLAFFPNXVJANFR-UHFFFAOYSA-N | |
2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one | |
96253 | |
90+% |
Glass bottle | |
5g | |
90% min. (HPLC) | |
2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 7-deazaguanine, gpl8t5zo3m, unii-gpl8t5zo3m | |
C1=CNC2=C1C(=O)N=C(N2)N | |
150.14 | |
150.14 |
Chemical Identifiers
7355-55-7 | |
150.14 | |
2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 7-deazaguanine, gpl8t5zo3m, unii-gpl8t5zo3m | |
2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one |
C6H6N4O | |
OLAFFPNXVJANFR-UHFFFAOYSA-N | |
96253 | |
C1=CNC2=C1C(=O)N=C(N2)N |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning