missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 7-Chloroquinoline-4-boronic acid pinacol ester, 96%
$200.14 - $713.19
Chemical Identifiers
CAS | 871125-83-6 |
---|---|
Molecular Formula | C15H17BClNO2 |
Molecular Weight (g/mol) | 289.566 |
MDL Number | MFCD07783020 |
InChI Key | GZDUYRAAWRRVEY-UHFFFAOYSA-N |
Synonym | 7-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 7-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 7-chloroquinolin-4-yl boronic acid pinacol ester, 7-chloroquinoline-4-boronic acid pinacol ester, 7-chloroquinoline-4-boronic acid, pinacol ester, quinoline,7-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl |
PubChem CID | 16218076 |
IUPAC Name | 7-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=C3C=CC(=CC3=NC=C2)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5103703
|
Alfa Aesar™
H5103703 |
1g |
Each for $200.14
|
|
AAH5103706
|
Alfa Aesar™
H5103706 |
5g |
Each for $713.19
|
|
Chemical Identifiers
871125-83-6 | |
289.566 | |
GZDUYRAAWRRVEY-UHFFFAOYSA-N | |
16218076 | |
B1(OC(C(O1)(C)C)(C)C)C2=C3C=CC(=CC3=NC=C2)Cl |
C15H17BClNO2 | |
MFCD07783020 | |
7-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 7-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 7-chloroquinolin-4-yl boronic acid pinacol ester, 7-chloroquinoline-4-boronic acid pinacol ester, 7-chloroquinoline-4-boronic acid, pinacol ester, quinoline,7-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
7-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline |
Specifications
1g | |
C15H17BClNO2 | |
7-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 7-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 7-chloroquinolin-4-yl boronic acid pinacol ester, 7-chloroquinoline-4-boronic acid pinacol ester, 7-chloroquinoline-4-boronic acid, pinacol ester, quinoline,7-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
B1(OC(C(O1)(C)C)(C)C)C2=C3C=CC(=CC3=NC=C2)Cl | |
289.566 | |
289.56 | |
7-Chloroquinoline-4-boronic acid pinacol ester |
871125-83-6 | |
MFCD07783020 | |
GZDUYRAAWRRVEY-UHFFFAOYSA-N | |
7-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline | |
16218076 | |
96% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures