missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Chloroisatin, 97%

Manufacturer:  Alfa Aesar™ H5559406

 View more versions of this product

Catalog No. AAH5559406



Specifications

Specifications

7-Chloroisatin
187°C to 191°C
7477-63-6
MFCD00022796
MPLXQMMMGDYXIT-UHFFFAOYSA-N
7-chloro-1H-indole-2,3-dione
344135
Solid
Orange
5g
C8H4ClNO2
1h-indole-2,3-dione, 7-chloro
C1=CC2=C(C(=C1)Cl)NC(=O)C2=O
181.575
181.58
97%

Chemical Identifiers

7477-63-6
181.575
MPLXQMMMGDYXIT-UHFFFAOYSA-N
344135
C1=CC2=C(C(=C1)Cl)NC(=O)C2=O
C8H4ClNO2
MFCD00022796
1h-indole-2,3-dione, 7-chloro
7-chloro-1H-indole-2,3-dione
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents