Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Chloroheptanenitrile, 99%

Chemical Identifiers

Chemical Name or Material 7-Chloroheptanenitrile
CAS 22819-91-6
Molecular Formula Cl(CH2)6CN
Formula Weight 145.63
MDL Number MFCD00039504
UN Number UN3276
Beilstein 1747798
Synonym 7-Chloroheptanonitrile; 6-Chlorohexyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0399709

 
alfa aesar™
L0399709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0399718

 
alfa aesar™
L0399718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 145.63
Chemical Name or Material 7-Chloroheptanenitrile
Assay 99%
CAS 22819-91-6
Molecular Formula Cl(CH2)6CN
MDL Number MFCD00039504
UN Number UN3276
Beilstein 1747798
Synonym 7-Chloroheptanonitrile; 6-Chlorohexyl cyanide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Quantity 10g
Boiling Point 80°C (0.6mmHg)
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.