missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Chloro-6-nitro-4(3H)-quinazolinone, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC450790250
Specifications
7-Chloro-6-nitro-4(3H)-quinazolinone | |
25g | |
96% min. (HPLC) | |
4 1h-quinazolinone, 7-chloro-6-nitro, 6-nitro-7-chloro-4-hydroxyquinazoline, 7-chloro-4-hydroxy-6-nitroquinazoline, 7-chloro-6-nitro-3,4-dihydroquinazolin-4-one, 7-chloro-6-nitro-3h-quinazolin-4-one, 7-chloro-6-nitro-4-hydroxyquinazoline, 7-chloro-6-nitro-quinazolin-4-one, 7-chloro-6-nitroquinazolin-4 3h-one, 7-chloro-6-nitroquinazolin-4-ol, 7-chloro-6-nitroquinazolin-4-one | |
URDYTQYZXZKBQT-UHFFFAOYSA-N | |
7-chloro-6-nitro-1H-quinazolin-4-one | |
12441237 | |
97% |
Glass bottle | |
53449-14-2 | |
C8H4ClN3O3 | |
Solubility in water: .. Other solubilities: slightly soluble in dmf | |
C1=C2C(=CC(=C1[N+](=O)[O-])Cl)NC=NC2=O | |
225.59 | |
225.59 |
Chemical Identifiers
53449-14-2 | |
225.59 | |
4 1h-quinazolinone, 7-chloro-6-nitro, 6-nitro-7-chloro-4-hydroxyquinazoline, 7-chloro-4-hydroxy-6-nitroquinazoline, 7-chloro-6-nitro-3,4-dihydroquinazolin-4-one, 7-chloro-6-nitro-3h-quinazolin-4-one, 7-chloro-6-nitro-4-hydroxyquinazoline, 7-chloro-6-nitro-quinazolin-4-one, 7-chloro-6-nitroquinazolin-4 3h-one, 7-chloro-6-nitroquinazolin-4-ol, 7-chloro-6-nitroquinazolin-4-one | |
7-chloro-6-nitro-1H-quinazolin-4-one |
C8H4ClN3O3 | |
URDYTQYZXZKBQT-UHFFFAOYSA-N | |
12441237 | |
C1=C2C(=CC(=C1[N+](=O)[O-])Cl)NC=NC2=O |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact
GHS Signal Word: Danger