missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Chloro-6-azaindole, 97%

$93.54 - $808.42

Chemical Identifiers

CAS 357263-41-3
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.581
MDL Number MFCD03839905
InChI Key HOHKYYCVFMEBGG-UHFFFAOYSA-N
Synonym 1h-pyrrolo 2,3-c pyridine, 7-chloro
PubChem CID 11435048
IUPAC Name 7-chloro-1H-pyrrolo[2,3-c]pyridine
SMILES C1=CNC2=C1C=CN=C2Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH28946MD View Documents Alfa Aesar™
H28946MD
250mg Each for $93.54
 
AAH2894603 View Documents Alfa Aesar™
H2894603
1g Each for $245.91
 
AAH2894606 View Documents Alfa Aesar™
H2894606
5g Each for $808.42
 
Specifications

Chemical Identifiers

357263-41-3
152.581
HOHKYYCVFMEBGG-UHFFFAOYSA-N
11435048
C1=CNC2=C1C=CN=C2Cl
C7H5ClN2
MFCD03839905
1h-pyrrolo 2,3-c pyridine, 7-chloro
7-chloro-1H-pyrrolo[2,3-c]pyridine

Specifications

250mg
C7H5ClN2
1h-pyrrolo 2,3-c pyridine, 7-chloro
C1=CNC2=C1C=CN=C2Cl
152.581
152.58
7-Chloro-6-azaindole
357263-41-3
MFCD03839905
HOHKYYCVFMEBGG-UHFFFAOYSA-N
7-chloro-1H-pyrrolo[2,3-c]pyridine
11435048
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures