Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Chloro-4-hydroxyquinoline, 98+%

$44.15 - $121.74

Chemical Identifiers

Chemical Name or Material 7-Chloro-4-hydroxyquinoline
CAS 86-99-7
Molecular Formula C9H6ClNO
Formula Weight 179.61
MDL Number MFCD00006778
Beilstein 125356
Synonym 7-Chloro-4-quinolinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0711914

  SDS
alfa aesar™
L0711914
25g Each for $44.15

AAL0711922

  SDS
alfa aesar™
L0711922
100g Each for $121.74
Description & Specifications

Specifications

Formula Weight 179.61
Molecular Formula C9H6ClNO
MDL Number MFCD00006778
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 86-99-7
Chemical Name or Material 7-Chloro-4-hydroxyquinoline
Beilstein 125356
Assay ≥98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Synonym 7-Chloro-4-quinolinol
Quantity 25g
Melting Point 278° to 282°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.