Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Chloro-2H-1,4-benzothiazin-3(4H)-one, 97%

$42.34 - $133.65

Chemical Identifiers

Chemical Name or Material 7-Chloro-2H-1,4-benzothiazin-3(4H)-one
CAS 5333-05-1
Molecular Formula C8H6ClNOS
Formula Weight 199.66
MDL Number MFCD00127344
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2715703

  SDS
alfa aesar™
H2715703
1g Each for $42.34

AAH2715706

  SDS
alfa aesar™
H2715706
5g Each for $133.65
Description & Specifications

Specifications

Formula Weight 199.66
Chemical Name or Material 7-Chloro-2H-1,4-benzothiazin-3(4H)-one
Assay 97%
Molecular Formula C8H6ClNOS
MDL Number MFCD00127344
CAS 5333-05-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 211° to 213°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.