Please login to your online account to display your discounted pricing

7-Chloro-1-ethyl-6-fluoro-4-oxohydroquinoline-3-carboxylic acid, 98%, Acros Organics

$97.38 - $97.38

Chemical Identifiers

Chemical Name or Material 7-Chloro-1-ethyl-6-fluoro-4-oxohydroquinoline-3-carboxylic acid
Name Note 98%
CAS 68077-26-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H9ClFNO3
Formula Weight 269.66
MDL Number MFCD00792459
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC335420050

  SDS
thermo fisher scientific
335420050
Each for $97.38
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 269.66
Quantity 5g
Color White
CAS Max % 100.0
Chemical Name or Material 7-Chloro-1-ethyl-6-fluoro-4-oxohydroquinoline-3-carboxylic acid
CAS Min % 97.5
Name Note 98%
CAS 68077-26-9
Assay Percent Range 98%
MDL Number MFCD00792459
Molecular Formula C12H9ClFNO3
Infrared Spectrum Authentic