Learn More
7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6345603
Description
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride | |
C9H11Cl2N | |
1 g | |
OGIAIXMUSSACDB-UHFFFAOYSA-N | |
7-chloro-1,2,3,4-tetrahydroisoquinoline;hydrochloride | |
12595070 | |
98% |
73075-45-3 | |
MFCD08461076 | |
7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-chloro-1,2,3,4-tetrahydroisoquinolinehydrochloride, isoquinoline, 7-chloro-1,2,3,4-tetrahydro-, hydrochloride, 7-chloro-1,2,3,4-tetrahydro-isoquinoline hcl, 7-chhloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochlor, 7-chloro-1,2,3,4-tetrahydroisoquinoline-hydrogen chloride 1/1 | |
C1CNCC2=C1C=CC(=C2)Cl.Cl | |
204.094 | |
204.1 | |
Solid |
Chemical Identifiers
73075-45-3 | |
204.094 | |
OGIAIXMUSSACDB-UHFFFAOYSA-N | |
12595070 | |
C1CNCC2=C1C=CC(=C2)Cl.Cl |
C9H11Cl2N | |
MFCD08461076 | |
7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-chloro-1,2,3,4-tetrahydroisoquinolinehydrochloride, isoquinoline, 7-chloro-1,2,3,4-tetrahydro-, hydrochloride, 7-chloro-1,2,3,4-tetrahydro-isoquinoline hcl, 7-chhloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochlor, 7-chloro-1,2,3,4-tetrahydroisoquinoline-hydrogen chloride 1/1 | |
7-chloro-1,2,3,4-tetrahydroisoquinoline;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only