missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Bromoisoquinoline, TCI America™

$67.22 - $198.61

Chemical Identifiers

CAS 58794-09-5
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD07368661
InChI Key KABRXLINDSPGDF-UHFFFAOYSA-N
PubChem CID 12257441
IUPAC Name 7-bromoisoquinoline
SMILES C1=CC(=CC2=C1C=CN=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B5071200MG View Documents TCI America
B5071200MG
200mg Each for $67.22
 
B50711G View Documents TCI America
B50711G
1g Each for $198.61
 
Specifications

Chemical Identifiers

58794-09-5
208.058
KABRXLINDSPGDF-UHFFFAOYSA-N
7-bromoisoquinoline
C9H6BrN
MFCD07368661
12257441
C1=CC(=CC2=C1C=CN=C2)Br

Specifications

Undesignated
200mg
C9H6BrN
KABRXLINDSPGDF-UHFFFAOYSA-N
7-bromoisoquinoline
12257441
Crystal-Powder at 20°C
7-Bromoisoquinoline
71°C
58794-09-5
MFCD07368661
C1=CC(=CC2=C1C=CN=C2)Br
208.058
208.06
≥96.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No