Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Bromoisoquinoline N-oxide, 98%

Chemical Identifiers

Chemical Name or Material 7-Bromoisoquinoline N-oxide
Molecular Formula C9H6BrNO
Formula Weight 224.06
MDL Number MFCD09953462
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2589906

  SDS
alfa aesar™
H2589906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2589914

  SDS
alfa aesar™
H2589914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2589903

  SDS
alfa aesar™
H2589903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 7-Bromoisoquinoline N-oxide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C9H6BrNO
Formula Weight 224.06
MDL Number MFCD09953462
Melting Point 181° to 188°C
Quantity 5g

Applications
7-Bromoisoquinoline N-oxide is the reactant in the direct regioselective phosphonation of heteroaryl N-oxides with H-phosphonates

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.