Learn More
7-Bromoisatin, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3284309
Description
7-bromoisatin was treated with excess PhMgBr to afford tertiary alcohol 9 in 96 % yield. Isatin oxime triflates undergo a facile fragmentation which is promoted by DBU, to form anthranilonitriles after hydrolytic work-up.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications7-bromoisatin was treated with excess PhMgBr to afford tertiary alcohol 9 in 96 % yield. Isatin oxime triflates undergo a facile fragmentation which is promoted by DBU, to form anthranilonitriles after hydrolytic work-up.
Solubility
Slightly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
7-Bromoisatin | |
195°C to 199°C | |
MFCD00774354 | |
7-bromoisatin, 7-bromoindoline-2,3-dione, 7-bromo-2,3-dioxoindoline, 1h-indole-2,3-dione, 7-bromo, 7-bromoindole-2,3-dione, 7-bromo-2,3-dihydro-1h-indole-2,3-dione, 7-bromoindole-1h-2,3-dione, 7-bromo-2,3-indolinedione, 7-bromo isatin, 7-bromo-isatin | |
OCVKSIWBTJCXPV-UHFFFAOYSA-N | |
7-bromo-1H-indole-2,3-dione | |
2302353 | |
97% |
20780-74-9 | |
C8H4BrNO2 | |
10 g | |
Slightly soluble. | |
C1=CC2=C(C(=C1)Br)NC(=O)C2=O | |
226.029 | |
226.03 |
Chemical Identifiers
20780-74-9 | |
226.029 | |
OCVKSIWBTJCXPV-UHFFFAOYSA-N | |
2302353 | |
C1=CC2=C(C(=C1)Br)NC(=O)C2=O |
C8H4BrNO2 | |
MFCD00774354 | |
7-bromoisatin, 7-bromoindoline-2,3-dione, 7-bromo-2,3-dioxoindoline, 1h-indole-2,3-dione, 7-bromo, 7-bromoindole-2,3-dione, 7-bromo-2,3-dihydro-1h-indole-2,3-dione, 7-bromoindole-1h-2,3-dione, 7-bromo-2,3-indolinedione, 7-bromo isatin, 7-bromo-isatin | |
7-bromo-1H-indole-2,3-dione |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only