Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Bromoindole, 98%

$372.50 - $372.50

Chemical Identifiers

Chemical Name or Material 7-Bromoindole
CAS 51417-51-7
Molecular Formula C8H6BrN
Formula Weight 196.05
MDL Number MFCD00799492
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4442206

 
alfa aesar™
4442206
5g Each for $372.50
Description & Specifications

Specifications

Formula Weight 196.05
Chemical Name or Material 7-Bromoindole
Melting Point 42° to 44°C
Quantity 5g
Sensitivity Light Sensitive
Physical Form Powder
Assay 98%
Molecular Formula C8H6BrN
MDL Number MFCD00799492
CAS 51417-51-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.