missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Bromoindole, 98%

Manufacturer:  Alfa Aesar™ 4442206

 View more versions of this product

Catalog No. AA4442206



Specifications

Specifications

7-Bromoindole
42°C to 44°C
51417-51-7
MFCD00799492
RDSVSEFWZUWZHW-UHFFFAOYSA-N
7-bromo-1H-indole
2757020
Powder
Light sensitive
5g
C8H6BrN
1h-indole, 7-bromo
C1=CC2=C(C(=C1)Br)NC=C2
196.047
196.05
98%

Chemical Identifiers

51417-51-7
196.047
RDSVSEFWZUWZHW-UHFFFAOYSA-N
2757020
C1=CC2=C(C(=C1)Br)NC=C2
C8H6BrN
MFCD00799492
1h-indole, 7-bromo
7-bromo-1H-indole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

Documents