missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Bromoindole, 98%

Manufacturer:  Alfa Aesar™ 4442203

 View more versions of this product

Catalog No. AA4442203



Specifications

Specifications

7-Bromoindole
42°C to 44°C
51417-51-7
MFCD00799492
RDSVSEFWZUWZHW-UHFFFAOYSA-N
7-bromo-1H-indole
2757020
Powder
Light Sensitive
1g
C8H6BrN
7-bromoindole, 1h-indole, 7-bromo, 7-bromo-indole, 7-bromindol, 7-bromoindol, 7-brom-1h-indole, pubchem7318, acmc-209ktm, 7-bromoindole, ksc269i1f
C1=CC2=C(C(=C1)Br)NC=C2
196.047
196.05
98%

Chemical Identifiers

51417-51-7
196.047
RDSVSEFWZUWZHW-UHFFFAOYSA-N
2757020
C1=CC2=C(C(=C1)Br)NC=C2
C8H6BrN
MFCD00799492
7-bromoindole, 1h-indole, 7-bromo, 7-bromo-indole, 7-bromindol, 7-bromoindol, 7-brom-1h-indole, pubchem7318, acmc-209ktm, 7-bromoindole, ksc269i1f
7-bromo-1H-indole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents