Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Bromoheptanenitrile, 98%

Chemical Identifiers

Chemical Name or Material 7-Bromoheptanenitrile
CAS 20965-27-9
Molecular Formula Br(CH2)6CN
Formula Weight 190.09
MDL Number MFCD00001982
UN Number UN3276
Beilstein 1748070
Synonym 7-Bromoheptanonitrile; 6-Bromohexyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0506806

  SDS
alfa aesar™
L0506806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0506814

  SDS
alfa aesar™
L0506814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.09
MDL Number MFCD00001982
Molecular Formula Br(CH2)6CN
CAS 20965-27-9
Synonym 7-Bromoheptanonitrile; 6-Bromohexyl cyanide
Health Hazard 1 Danger
Assay 98%
TSCA TSCA
UN Number UN3276
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Beilstein 1748070
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Flash Point >110°C (230°F)
Boiling Point 114° to 115°C (4mmHg)
Quantity 5g
Chemical Name or Material 7-Bromoheptanenitrile
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.