Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Bromoacetyl-3,4-dihydro-1,5-benzodioxepin, 97%

Chemical Identifiers

Chemical Name or Material 7-Bromoacetyl-3,4-dihydro-1,5-benzodioxepin
CAS 35970-34-4
Molecular Formula C11H11BrO3
Formula Weight 271.11
MDL Number MFCD00218462
UN Number UN1759
Synonym 3,4-(Trimethylenedioxy)phenacyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2731403

  SDS
alfa aesar™
H2731403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2731406

  SDS
alfa aesar™
H2731406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 271.11
Chemical Name or Material 7-Bromoacetyl-3,4-dihydro-1,5-benzodioxepin
Assay 97%
Molecular Formula C11H11BrO3
MDL Number MFCD00218462
CAS 35970-34-4
UN Number UN1759
Melting Point 74° to 78°C
Synonym 3,4-(Trimethylenedioxy)phenacyl bromide
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.