missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Bromo-5-chloro-8-hydroxyquinoline 98.0+%, TCI America™
$173.56 - $484.70
Chemical Identifiers
CAS | 7640-33-7 |
---|---|
Molecular Formula | C9H5BrClNO |
Molecular Weight (g/mol) | 258.499 |
MDL Number | MFCD00044601 |
InChI Key | ADJQQSUBKRASQN-UHFFFAOYSA-N |
PubChem CID | 82095 |
IUPAC Name | 7-bromo-5-chloroquinolin-8-ol |
SMILES | C1=CC2=C(C(=C(C=C2Cl)Br)O)N=C1 |
Chemical Identifiers
7640-33-7 | |
258.499 | |
ADJQQSUBKRASQN-UHFFFAOYSA-N | |
7-bromo-5-chloroquinolin-8-ol |
C9H5BrClNO | |
MFCD00044601 | |
82095 | |
C1=CC2=C(C(=C(C=C2Cl)Br)O)N=C1 |
Specifications
Undesignated | |
1g | |
C9H5BrClNO | |
ADJQQSUBKRASQN-UHFFFAOYSA-N | |
7-bromo-5-chloroquinolin-8-ol | |
82095 | |
Crystal-Powder at 20°C | |
7-Bromo-5-chloro-8-hydroxyquinoline |
181°C | |
7640-33-7 | |
MFCD00044601 | |
C1=CC2=C(C(=C(C=C2Cl)Br)O)N=C1 | |
258.499 | |
258.50 | |
≥98.0% (GC,T) |
Safety and Handling
RTECSNumber : VC4585000
TSCA : No