missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Bromo-4-fluoroindole, 97%

$88.24 - $1,051.11

Chemical Identifiers

CAS 292636-09-0
Molecular Formula C8H5BrFN
Molecular Weight (g/mol) 214.037
MDL Number MFCD04037874
InChI Key PRXUTIXCOBFMMM-UHFFFAOYSA-N
Synonym 1h-indole, 7-bromo-4-fluoro
PubChem CID 4342956
IUPAC Name 7-bromo-4-fluoro-1H-indole
SMILES C1=CC(=C2C(=C1F)C=CN2)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54387MD View Documents Alfa Aesar™
H54387MD
250mg Each for $88.24
 
AAH5438703 View Documents Alfa Aesar™
H5438703
1g Each for $261.97
 
AAH5438706 View Documents Alfa Aesar™
H5438706
5g Each for $1,051.11
 
Specifications

Chemical Identifiers

292636-09-0
214.037
PRXUTIXCOBFMMM-UHFFFAOYSA-N
4342956
C1=CC(=C2C(=C1F)C=CN2)Br
C8H5BrFN
MFCD04037874
1h-indole, 7-bromo-4-fluoro
7-bromo-4-fluoro-1H-indole

Specifications

Air sensitive
292636-09-0
MFCD04037874
PRXUTIXCOBFMMM-UHFFFAOYSA-N
7-bromo-4-fluoro-1H-indole
4342956
97%
250mg
C8H5BrFN
1h-indole, 7-bromo-4-fluoro
C1=CC(=C2C(=C1F)C=CN2)Br
214.037
214.04
7-Bromo-4-fluoroindole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Argon