missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Bromo-4-fluoroindole, 97%

Manufacturer:  Alfa Aesar™ H5438706

 View more versions of this product

Catalog No. AAH5438706



Specifications

Specifications

7-Bromo-4-fluoroindole
292636-09-0
MFCD04037874
PRXUTIXCOBFMMM-UHFFFAOYSA-N
7-bromo-4-fluoro-1H-indole
4342956
97%
5g
C8H5BrFN
1h-indole, 7-bromo-4-fluoro
C1=CC(=C2C(=C1F)C=CN2)Br
214.037
214.04

Chemical Identifiers

292636-09-0
214.037
PRXUTIXCOBFMMM-UHFFFAOYSA-N
4342956
C1=CC(=C2C(=C1F)C=CN2)Br
C8H5BrFN
MFCD04037874
1h-indole, 7-bromo-4-fluoro
7-bromo-4-fluoro-1H-indole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents