Please login to your online account to display your discounted pricing

7-Bromo-3,4-dihydronaphthalen-1(2H)-one, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 7-Bromo-3, 4-dihydronaphthalen-1(2H)-one
Assay Percent Range 97%
Molecular Formula C10H9BrO
Formula Weight 225.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC448680100

  SDS
thermo fisher scientific
AC448680100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC448680500

  SDS
thermo fisher scientific
AC448680500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 7-Bromo-3, 4-dihydronaphthalen-1(2H)-one
CAS 32281-97-3
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C10H9BrO
Formula Weight 225.08
Quantity 10g