Please login to your online account to display your discounted pricing

7-Bromo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC60810DE

 View more versions of this product

Catalog No. CC60810DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 215.05
Quantity 5g
Assay 97%
Molecular Formula C7H7BrN2O
Assay Percent Range 97%
Packaging Amber Glass Bottle
CAS 34950-82-8
Chemical Name or Material 7-Bromo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine

Specifications

Chemical Name or Material 7-Bromo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H7BrN2O
Formula Weight 215.05