missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Bromo-3,4-dihydro-2H-1,4-benzoxazine, 97%

Manufacturer:  Alfa Aesar™ H61071MD

 View more versions of this product

Catalog No. AAH61071MD



Specifications

Specifications

250mg
C8H8BrNO
2h-1,4-benzoxazine, 7-bromo-3,4-dihydro, 2h-1,4-benzoxazine,7-bromo-3,4-dihydro, 7-bromo-3,4-dihydro-2h-benzo 1,4 oxazine, 7-bromo-3,4-dihydro-2h-benzo b 1,4 oxazin, 7-bromo-3,4-dihydro-2h-benzo b 1,4 oxazine, acmc-209yv4, pubchem22676
C1COC2=C(N1)C=CC(=C2)Br
214.062
214.06
105679-22-9
MFCD09056750
JLZUUGCTPRPFKZ-UHFFFAOYSA-N
7-bromo-3,4-dihydro-2H-1,4-benzoxazine
18008960
97%

Chemical Identifiers

105679-22-9
214.062
JLZUUGCTPRPFKZ-UHFFFAOYSA-N
18008960
C1COC2=C(N1)C=CC(=C2)Br
C8H8BrNO
MFCD09056750
2h-1,4-benzoxazine, 7-bromo-3,4-dihydro, 2h-1,4-benzoxazine,7-bromo-3,4-dihydro, 7-bromo-3,4-dihydro-2h-benzo 1,4 oxazine, 7-bromo-3,4-dihydro-2h-benzo b 1,4 oxazin, 7-bromo-3,4-dihydro-2h-benzo b 1,4 oxazine, acmc-209yv4, pubchem22676
7-bromo-3,4-dihydro-2H-1,4-benzoxazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents