Learn More
7-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H61021MD
Description
7-Bromo-3,4-dihydro-2H-1,4-benzothiazine used in many areas of medicine. They are used in industries as dyes and as antioxidants of rubber and natural elastomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications7-Bromo-3,4-dihydro-2H-1,4-benzothiazine used in many areas of medicine. They are used in industries as dyes and as antioxidants of rubber and natural elastomers.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
7-Bromo-3,4-dihydro-2H-1,4-benzothiazine | |
C8H8BrNS | |
250 mg | |
QWWYGAWGUCQKDL-UHFFFAOYSA-N | |
7-bromo-3,4-dihydro-2H-1,4-benzothiazine | |
83823811 | |
97% |
193414-60-7 | |
MFCD11847754 | |
7-bromo-3,4-dihydro-2h-benzo b 1,4 thiazine | |
BrC1=CC2=C(NCCS2)C=C1 | |
230.12 | |
230.12 |
Chemical Identifiers
193414-60-7 | |
230.12 | |
QWWYGAWGUCQKDL-UHFFFAOYSA-N | |
83823811 | |
BrC1=CC2=C(NCCS2)C=C1 |
C8H8BrNS | |
MFCD11847754 | |
7-bromo-3,4-dihydro-2h-benzo b 1,4 thiazine | |
7-bromo-3,4-dihydro-2H-1,4-benzothiazine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only