Learn More
7-Bromo-1-hydroxyisoquinoline, 97%, Thermo Scientific Chemicals
$164.92 - $627.19
Chemical Identifiers
CAS | 223671-15-6 |
---|---|
Molecular Formula | C9H6BrNO |
Molecular Weight (g/mol) | 224.057 |
MDL Number | MFCD02093963 |
InChI Key | DSOKREQUHLPVFR-UHFFFAOYSA-N |
PubChem CID | 11276133 |
IUPAC Name | 7-bromo-2H-isoquinolin-1-one |
SMILES | C1=CC(=CC2=C1C=CNC2=O)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1780003
|
Thermo Scientific Chemicals
L1780003 |
1 g |
Each for $164.92
|
|
|||||
AAL1780006
|
Thermo Scientific Chemicals
L1780006 |
5 g |
Each for $627.19
|
|
|||||
Description
7-Bromo-1-hydroxyisoquinoline acts as a pharmaceutical intermediate. It is also used to prepare 1-amino-4-bromoisoquinoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications7-Bromo-1-hydroxyisoquinoline acts as a pharmaceutical intermediate. It is also used to prepare 1-amino-4-bromoisoquinoline.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
223671-15-6 | |
224.057 | |
DSOKREQUHLPVFR-UHFFFAOYSA-N | |
7-bromo-2H-isoquinolin-1-one |
C9H6BrNO | |
MFCD02093963 | |
11276133 | |
C1=CC(=CC2=C1C=CNC2=O)Br |
Specifications
223671-15-6 | |
C9H6BrNO | |
1 g | |
7-bromoisoquinolin-1-ol, 7-bromo-1-hydroxyisoquinoline, 7-bromoisoquinolin-1 2h-one, 1 2h-isoquinolinone, 7-bromo, 1 2h-isoquinolinone,7-bromo, 7-bromo-1,2-dihydroisoquinolin-1-one, 7-bromoisoquinolone, 7-bromoisoquinolinone, 7-bromoisocarbostyril, 7-bromo-1-isoquinolinol | |
DSOKREQUHLPVFR-UHFFFAOYSA-N | |
7-bromo-2H-isoquinolin-1-one | |
11276133 | |
≥98% |
248°C to 250°C | |
MFCD02093963 | |
8405244 | |
Slightly soluble in water. | |
C1=CC(=CC2=C1C=CNC2=O)Br | |
224.057 | |
224.06 | |
7-Bromo-1-hydroxyisoquinoline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only