missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Bromo-1-heptanol, 96%

Manufacturer:  Alfa Aesar™ H54762MD

 View more versions of this product

Catalog No. AAH54762MD



Description

Description

Applications
7-Bromo-1-heptanol was used in the synthesis of (9Z,12E)-[1-13C]-octadeca-9,12-dienoic acid and(9Z,12Z,15E)-[1-13C]-octadeca-9,12,15-trienoic acid. It can be used to prepare N-substituted [(carboxymethyl)phenoxy]ethyl benzoxazinones as PPARγ agonists and antidiabetic (type 2) agents. It can also be used to prepare thiochroman and chroman derivatives as pure antiestrogens.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Specifications

7-Bromo-1-heptanol
111°C to 112°C (4mmHg)
10160-24-4
Br(CH2)7OH
1-heptanol, 7-bromo
C(CCCO)CCCBr
195.1
195.1
113°C (235°F)
250mg
C7H15BrO
MFCD00041708
MMXRRNUXCHUHOE-UHFFFAOYSA-N
7-bromoheptan-1-ol
66284
96%

Chemical Identifiers

10160-24-4
195.1
MMXRRNUXCHUHOE-UHFFFAOYSA-N
66284
C(CCCO)CCCBr
C7H15BrO
MFCD00041708
1-heptanol, 7-bromo
7-bromoheptan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents