missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Bromo-1-heptanol, 96%

Manufacturer:  Alfa Aesar™ H5476206

 View more versions of this product

Catalog No. AAH5476206



Description

Description

Applications
7-Bromo-1-heptanol was used in the synthesis of (9Z,12E)-[1-13C]-octadeca-9,12-dienoic acid and(9Z,12Z,15E)-[1-13C]-octadeca-9,12,15-trienoic acid. It can be used to prepare N-substituted [(carboxymethyl)phenoxy]ethyl benzoxazinones as PPARγ agonists and antidiabetic (type 2) agents. It can also be used to prepare thiochroman and chroman derivatives as pure antiestrogens.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Specifications

7-Bromo-1-heptanol
113°C (235°F)
111°C to 112°C (4mmHg)
10160-24-4
Br(CH2)7OH
1-heptanol, 7-bromo
MMXRRNUXCHUHOE-UHFFFAOYSA-N
7-bromoheptan-1-ol
66284
96%
1.269
1.482
5g
C7H15BrO
MFCD00041708
Slightly soluble in water.
C(CCCO)CCCBr
195.1
195.1

Chemical Identifiers

10160-24-4
195.1
MMXRRNUXCHUHOE-UHFFFAOYSA-N
66284
C(CCCO)CCCBr
C7H15BrO
MFCD00041708
1-heptanol, 7-bromo
7-bromoheptan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents