Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Bromo-1,2,3,4-tetrahydrocyclopent[b]indole, 99%

$116.99 - $404.82

Chemical Identifiers

Chemical Name or Material 7-Bromo-1,2,3,4-tetrahydrocyclopent[b]indole
CAS 164736-47-4
Molecular Formula C11H10BrN
Formula Weight 236.11
MDL Number MFCD00239009
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5059203

  SDS
alfa aesar™
H5059203
1g Each for $404.82

AAH50592MD

  SDS
alfa aesar™
H50592MD
250mg Each for $116.99
Description & Specifications

Specifications

Formula Weight 236.11
Chemical Name or Material 7-Bromo-1,2,3,4-tetrahydrocyclopent[b]indole
Assay 99%
Molecular Formula C11H10BrN
MDL Number MFCD00239009
CAS 164736-47-4
Quantity 1g