Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Benzyloxyindole-3-carboxaldehyde, 97%

$92.04 - $297.08

Chemical Identifiers

Chemical Name or Material 7-Benzyloxyindole-3-carboxaldehyde
CAS 92855-65-7
Molecular Formula C16H13NO2
Formula Weight 251.28
MDL Number MFCD00055979
Synonym 7-Benzyloxy-3-formylindole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2877703

 
alfa aesar™
H2877703
1g Each for $297.08

AAH28777MD

 
alfa aesar™
H28777MD
250mg Each for $92.04
Description & Specifications

Specifications

Formula Weight 251.28
Chemical Name or Material 7-Benzyloxyindole-3-carboxaldehyde
Assay 97%
Molecular Formula C16H13NO2
MDL Number MFCD00055979
CAS 92855-65-7
Synonym 7-Benzyloxy-3-formylindole
Melting Point 152° to 159°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.