Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Benzoylheptanoic acid, 98+%

$77.59 - $266.70

Chemical Identifiers

Chemical Name or Material 7-Benzoylheptanoic acid
CAS 66147-75-9
Molecular Formula C14H18O3
Formula Weight 234.3
MDL Number MFCD00039609
Beilstein 2649061
Synonym 8-Oxo-8-phenyloctanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0839503

  SDS
alfa aesar™
L0839503
1g Each for $77.59

AAL0839506

  SDS
alfa aesar™
L0839506
5g Each for $266.70
Description & Specifications

Specifications

Formula Weight 234.3
Chemical Name or Material 7-Benzoylheptanoic acid
Assay ≥98%
MDL Number MFCD00039609
CAS 66147-75-9
Beilstein 2649061
Synonym 8-Oxo-8-phenyloctanoic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C14H18O3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 80° to 82°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.