Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Azaindole-4-carboxylic acid, 97%

Chemical Identifiers

Chemical Name or Material 7-Azaindole-4-carboxylic acid
CAS 479553-01-0
Formula Weight 162.15
MDL Number MFCD08272236
Synonym 1H-Pyrrolo[2,3-b]pyridine-4-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3313903

  SDS
alfa aesar™
H3313903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH33139MD

  SDS
alfa aesar™
H33139MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.15
Melting Point >330°C
MDL Number MFCD08272236
Color White to cream
Physical Form Solid
Quantity 1g
Health Hazard 1 Warning
Chemical Name or Material 7-Azaindole-4-carboxylic acid
CAS 479553-01-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Synonym 1H-Pyrrolo[2,3-b]pyridine-4-carboxylic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.