missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%, Thermo Scientific™

Chemical Identifiers

CAS 156311-83-0
Molecular Formula C17H27F6N7OP2
Molecular Weight (g/mol) 521.39
MDL Number MFCD03703417
InChI Key CBZAHNDHLWAZQC-UHFFFAOYSA-N
Synonym 3-hydroxy-3h-1,2,3-triazolo 4,5-b pyridinato-o tri-1-pyrrolidinylphosphonium hexafluorophosphate, 3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxy tripyrrolidino-phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxy-tris-pyrrolidino phosphonium hexafluorophosphate, ksc491e1h, pubchem12746, pyaop, pyaop reagent
PubChem CID 11038641
IUPAC Name tripyrrolidin-1-yl(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.C1CCN(C1)[P+](ON1N=NC2=CC=CN=C12)(N1CCCC1)N1CCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC440610010
SDS View Documents Promotion Details
Thermo Fisher Scientific
440610010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC440610050
View Documents Promotion Details
Thermo Fisher Scientific
440610050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Coupling reagent
Specifications

Chemical Identifiers

156311-83-0
521.39
CBZAHNDHLWAZQC-UHFFFAOYSA-N
11038641
F[P-](F)(F)(F)(F)F.C1CCN(C1)[P+](ON1N=NC2=CC=CN=C12)(N1CCCC1)N1CCCC1
C17H27F6N7OP2
MFCD03703417
3-hydroxy-3h-1,2,3-triazolo 4,5-b pyridinato-o tri-1-pyrrolidinylphosphonium hexafluorophosphate, 3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxy tripyrrolidino-phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxy-tris-pyrrolidino phosphonium hexafluorophosphate, ksc491e1h, pubchem12746, pyaop, pyaop reagent
tripyrrolidin-1-yl(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate

Specifications

156311-83-0
100.0
153°C to 155°C
98%
Authentic
Glass bottle
3-hydroxy-3h-1,2,3-triazolo 4,5-b pyridinato-o tri-1-pyrrolidinylphosphonium hexafluorophosphate, 3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxy tripyrrolidino-phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxy-tris-pyrrolidino phosphonium hexafluorophosphate, ksc491e1h, pubchem12746, pyaop, pyaop reagent
F[P-](F)(F)(F)(F)F.C1CCN(C1)[P+](ON1N=NC2=CC=CN=C12)(N1CCCC1)N1CCCC1
521.39
521.39
98%
97.5
White
38.18 to 40.14% C, 18.33 to 19.28% N
C17H27F6N7OP2
MFCD03703417
1g
CBZAHNDHLWAZQC-UHFFFAOYSA-N
tripyrrolidin-1-yl(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate
11038641
Powder or Crystalline Powder
(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Product Certifications

RUO – Research Use Only