missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H2660906
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)Phosphonium Hexafluorophosphate | |
∽180°C (decomposition) | |
MFCD09263289 | |
3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxytris dimethylamino phosph, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, acmc-20alxg, tris dimethylamino 3h-1,2,3-triazolo 4,5-b pyridin-3-yloxy phosphonium hexafluorophosphate, ksc173q2f, 7-azabenzotriazol-1-yloxytris dimethyamino phosphonium hexafluorosphate, 7-azabenzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 98+% | |
CN(C)[P+](N(C)C)(N(C)C)ON1C2=C(C=CC=N2)N=N1.F[P-](F)(F)(F)(F)F | |
443.275 | |
443.27 |
156311-85-2 | |
C11H21F6N7OP2 | |
5 g | |
RQBNNDQCKMIUQJ-UHFFFAOYSA-N | |
tris(dimethylamino)-(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate | |
21894514 | |
≥98% |
Chemical Identifiers
156311-85-2 | |
443.275 | |
RQBNNDQCKMIUQJ-UHFFFAOYSA-N | |
21894514 | |
CN(C)[P+](N(C)C)(N(C)C)ON1C2=C(C=CC=N2)N=N1.F[P-](F)(F)(F)(F)F |
C11H21F6N7OP2 | |
MFCD09263289 | |
3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxytris dimethylamino phosph, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, acmc-20alxg, tris dimethylamino 3h-1,2,3-triazolo 4,5-b pyridin-3-yloxy phosphonium hexafluorophosphate, ksc173q2f, 7-azabenzotriazol-1-yloxytris dimethyamino phosphonium hexafluorosphate, 7-azabenzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 98+% | |
tris(dimethylamino)-(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold