missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Anilino-1-naphthol-3-sulfonic Acid 95.0+%, TCI America™

Manufacturer:  TCI America P013725G

 View more versions of this product

Catalog No. P013725G



Specifications

Specifications

7-Anilino-1-naphthol-3-sulfonic Acid
25g
C16H13NO4S
4-Hydroxy-6-(phenylamino)-2-naphthalenesulfonic Acid, Phenyl gamma-Acid
C1=CC=C(C=C1)NC2=CC3=C(C=C(C=C3C=C2)S(=O)(=O)O)O
315.343
315.34
≥95.0% (HPLC,N)
Undesignated
119-19-7
MFCD00021509
QEAYLNJEDDOYNQ-UHFFFAOYSA-N
6-anilino-4-hydroxynaphthalene-2-sulfonic acid
67062
Crystal-Powder at 20°C

Chemical Identifiers

119-19-7
315.343
QEAYLNJEDDOYNQ-UHFFFAOYSA-N
67062
C1=CC=C(C=C1)NC2=CC3=C(C=C(C=C3C=C2)S(=O)(=O)O)O
C16H13NO4S
MFCD00021509
4-Hydroxy-6-(phenylamino)-2-naphthalenesulfonic Acid, Phenyl gamma-Acid
6-anilino-4-hydroxynaphthalene-2-sulfonic acid
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1105

TSCA : No

Documents