Please login to your online account to display your discounted pricing

7-Aminothieno[2,3-b]pyrazine-6-carboxylic acid, ≥90%, Maybridge

Chemical Identifiers

Chemical Name or Material 7-Aminothieno[2, 3-b]pyrazine-6-carboxylic acid
CAS 56881-31-3
CAS Min % 90.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C7H5N3O2S
Formula Weight 195.2
MDL Number MFCD08271964
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07475CB

  SDS
fisher scientific company
MO07475CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.2
Quantity 250mg
Assay 90%
CAS Min % 90.0
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD08271964
Chemical Name or Material 7-Aminothieno[2, 3-b]pyrazine-6-carboxylic acid
Molecular Formula C7H5N3O2S
Packaging Amber Glass Bottle
Assay Percent Range 90%
CAS 56881-31-3
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.