missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Aminoindole, 97%

$169.64 - $473.43

Chemical Identifiers

CAS 5192-04-1
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD02093960
InChI Key WTFWZOSMUGZKNZ-UHFFFAOYSA-N
Synonym 7-aminoindole, 7-amino-1h-indole, 1h-indol-7-ylamine, 7-indolamine, indole-7-ylamine, 7-indolylamine, 7-aminoindol, acmc-1ay6v, indol-7-yl amine, 7-aminoindole hplc
PubChem CID 111634
ChEBI CHEBI:33068
IUPAC Name 1H-indol-7-amine
SMILES C1=CC2=C(C(=C1)N)NC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL17794MC View Documents Alfa Aesar™
L17794MC
100mg Each for $169.64
 
AAL17794ME View Documents Alfa Aesar™
L17794ME
500mg Each for $473.43
 
Specifications

Chemical Identifiers

5192-04-1
132.17
WTFWZOSMUGZKNZ-UHFFFAOYSA-N
111634
1H-indol-7-amine
C8H8N2
MFCD02093960
7-aminoindole, 7-amino-1h-indole, 1h-indol-7-ylamine, 7-indolamine, indole-7-ylamine, 7-indolylamine, 7-aminoindol, acmc-1ay6v, indol-7-yl amine, 7-aminoindole hplc
CHEBI:33068
C1=CC2=C(C(=C1)N)NC=C2

Specifications

90°C to 91°C
5192-04-1
MFCD02093960
7-aminoindole, 7-amino-1h-indole, 1h-indol-7-ylamine, 7-indolamine, indole-7-ylamine, 7-indolylamine, 7-aminoindol, acmc-1ay6v, indol-7-yl amine, 7-aminoindole hplc
C1=CC2=C(C(=C1)N)NC=C2
132.17
CHEBI:33068
97%
100mg
C8H8N2
471787
WTFWZOSMUGZKNZ-UHFFFAOYSA-N
1H-indol-7-amine
111634
132.17
7-Aminoindole
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : −30° to −10°C