Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Aminodesacetoxycephalosporanic acid, 98%

$45.72 - $357.11

Chemical Identifiers

Chemical Name or Material 7-Aminodesacetoxycephalosporanic acid
CAS 22252-43-3
Molecular Formula C8H10N2O3S
Formula Weight 214.24
MDL Number MFCD00151456
Beilstein 5273616
Synonym 7-ADCA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0341706

  SDS
alfa aesar™
L0341706
5g Each for $357.11

AAL0341703

  SDS
alfa aesar™
L0341703
1g Each for $113.45

AAL03417MD

  SDS
alfa aesar™
L03417MD
250mg Each for $45.72
Description & Specifications

Specifications

Formula Weight 214.24
CAS 22252-43-3
Melting Point ≈234°C
Assay 98%
Quantity 5g
Molecular Formula C8H10N2O3S
MDL Number MFCD00151456
Beilstein 5273616
Synonym 7-ADCA
Recommended Storage 2° to 8°C
Chemical Name or Material 7-Aminodesacetoxycephalosporanic acid

Applications
7-Aminodesacetoxycephalosporanic acid is the Cephalexin metabolite. It is a Potent inhibitor of bacterial cell-wall enzyme. It is used in the synthesis of cephalosporins and for bioconversion studies.

Solubility
Soluble in water (partly), 1M ammonium hydroxide (50 mg/ml), and DMSO (Sparingly).

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Safety and Handling

2° to 8°C